Please login to your online account to display your discounted pricing

anti-MALT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056158

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056158

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity The antibody recognizes both human MALT1 isoforms a and b. Tranlocations between the MALT1 gene and other genes, can result in MALT1 chimeric (fusion) proteins of various sizes.
Gene ID (Entrez) 10892
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Full-length recombinant human MALT1/Paracaspase protein was used as immunogen.
Host Species Rabbit
Antigen MALT1
Regulatory Status RUO
Quantity 0.05mL
Format Antiserum