Please login to your online account to display your discounted pricing

anti-Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human JARID1B protein (between residues 800-850) [UniProt Q9UGL1]
Antigen Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP197310

 
novus biologicals canada
NBP197310
Unlabeled 0.1mL Each for $329.00

NBP197310SS

 
novus biologicals canada
NBP197310SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human JARID1B protein (between residues 800-850) [UniProt Q9UGL1]
Antigen Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9UGL1
Gene ID (Entrez) 10765
Formulation PBS, pH 7.4
Concentration 0.828mg/mL