Please login to your online account to display your discounted pricing

anti-Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164]
Antigen Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149602SS

 
novus biologicals canada
NBP149602SS
Unlabeled 0.025mL Each for $109.00

NBP149602

 
novus biologicals canada
NBP149602
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164]
Antigen Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene Accession No. O75164
Gene ID (Entrez) 9682
Formulation PBS, 30% glycerol
Concentration 0.53mg/mL