Please login to your online account to display your discounted pricing

anti-Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149602SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149602SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O75164
Concentration 0.53mg/mL
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Formulation PBS, 30% glycerol
Immunogen A genomic peptide made to an internal region of the human JMJD2A protein (within residues 320-460). [Swiss-Prot O75164]
Gene ID (Entrez) 9682
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Antigen Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Quantity 0.025mL