Please login to your online account to display your discounted pricing

anti-Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD1A protein (within residues 250-400). [Swiss-Prot Q9Y4C1]
Antigen Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149601

 
novus biologicals canada
NBP149601
Unlabeled 0.1mL Each for $329.00

NBP149601SS

 
novus biologicals canada
NBP149601SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD1A protein (within residues 250-400). [Swiss-Prot Q9Y4C1]
Antigen Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human, mouse and primate. Immunogen sequence has 91% homology with rat.
Gene Accession No. Q9Y4C1
Gene ID (Entrez) 55818
Formulation PBS, 30% glycerol
Concentration 0.53mg/mL