Please login to your online account to display your discounted pricing

anti-Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149601SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149601SS

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Human, mouse and primate. Immunogen sequence has 91% homology with rat.
Concentration 0.53mg/mL
Regulatory Status RUO
Gene Accession No. Q9Y4C1
Gene ID (Entrez) 55818
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human JMJD1A protein (within residues 250-400). [Swiss-Prot Q9Y4C1]
Quantity 0.025mL
Antigen Lysine (K)-specific Demethylase 3A/KDM3A/JMJD1A