Please login to your online account to display your discounted pricing

anti-Lyl1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen 235 RSQPAPPADPDGSPGG 250
Antigen Lyl1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227188

 
novus biologicals canada
NBP227188
Unlabeled 0.1mg Each for $257.41

NBP227188SS

 
novus biologicals canada
NBP227188SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 235 RSQPAPPADPDGSPGG 250
Antigen Lyl1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Test Specificity Dog, Human, Orangutan, Wild pig.
Gene ID (Entrez) 4066