Please login to your online account to display your discounted pricing

anti-LYK5, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 300-350 of human STRAD (isoform 1) was used as the immunogen.
Antigen LYK5
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224700

 
novus biologicals canada
NBP224700
Unlabeled 0.1mg Each for $255.80

NBP224700SS

 
novus biologicals canada
NBP224700SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 300-350 of human STRAD (isoform 1) was used as the immunogen.
Antigen LYK5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used is 100% homologous in Yellow Fluorescent Protein (YFP) and Cyan Fluorescent Protein (CFP). The NCBI accession number for GFP is AAC78774.
Gene ID (Entrez) 92335
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL