Please login to your online account to display your discounted pricing

anti-LYK5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224700SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224700SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 92335
Test Specificity The amino acid sequence used is 100% homologous in Yellow Fluorescent Protein (YFP) and Cyan Fluorescent Protein (CFP). The NCBI accession number for GFP is AAC78774.
Applications Western Blotting
Regulatory Status RUO
Isotype IgG
Cross Reactivity Human
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Host Species Rabbit
Format Purified
Antigen LYK5
Quantity 0.025mg
Immunogen A portion of amino acids 300-350 of human STRAD (isoform 1) was used as the immunogen.