Please login to your online account to display your discounted pricing

anti-LYK5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224700

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224700

  • / Each
 

Description & Specifications

Specifications

Antigen LYK5
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human STRAD (isoform 1) was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 92335
Test Specificity The amino acid sequence used is 100% homologous in Yellow Fluorescent Protein (YFP) and Cyan Fluorescent Protein (CFP). The NCBI accession number for GFP is AAC78774.
Target Species Human