Please login to your online account to display your discounted pricing

anti-Ly-6G6F, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 66-100 of human G6D was used as immunogen
Antigen Ly-6G6F
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224668

 
novus biologicals canada
NBP224668
Unlabeled 0.1mg Each for $257.41

NBP224668SS

 
novus biologicals canada
NBP224668SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 66-100 of human G6D was used as immunogen
Antigen Ly-6G6F
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 95% homologous in mouse, horse, rat, rhesus monkey and dog, 90% homologous in cow, 80% homologous in platypus and 70% homologous in opossum.
Gene Accession No. Q5SQ64
Gene ID (Entrez) 259215
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL