anti-Ly-6G, PE, Clone: 1A8, EMD Millipore™
Rat Monoclonal Antibody
Manufacturer:  Millipore Sigma MABF1423
 View more versions of this product
Catalog No. MABF1423MI
Description
Description
Specifically detects Ly-6G Clone: 1A8 in Mouse samples, and it is validated for Flow Cytometry
The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes.
Specifications
Specifications
Ly-6G
1A8
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Purified
Rat
IgG2a, κ
100μg
Inflammation, Immunology
Primary
Murine
Flow Cytometry
PE
P35461
Ly6g
Proprietary immunogen
Purified by size-exclusion chromatography
RUO
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Monoclonal