Please login to your online account to display your discounted pricing

anti-LXR, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt No. Q13133]
Antigen LXR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB400157

 
novus biologicals canada
NB400157
Unlabeled 0.1mL Each for $362.00

NB400157SS

 
novus biologicals canada
NB400157SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt No. Q13133]
Antigen LXR
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, mouse, porcine and monkey. Immunogen sequence has 92% identity to rat and 90% identity to chicken.
Gene Accession No. Q13133
Gene ID (Entrez) 10062
Formulation Tris-glycine, 150mM NaCl
Concentration 1.04mg/mL