Please login to your online account to display your discounted pricing

anti-LTK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056425

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056425

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4058
Conjugate Unlabeled
Gene Accession No. P29376
Cross Reactivity Human
Format Purified
Immunogen A portion of amino acids 800-864 of human LTK were used as the immunogen.
Test Specificity The amino acid sequence used as immunogen has 100% identity in human isoforms 1 and 2, and 81% homologous in mouse isoforms A, B, C, and D, and rat. LTK is an 864 amino acid protein with an approximate molecular weight of 94 kD. Cross reacts with Human. Predicted to cross react with Mouse and Rat.
Concentration 0.5mg/mL
Quantity 0.1mg
Antigen LTK
Host Species Rabbit