Please login to your online account to display your discounted pricing

anti-LTK, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 800-864 of human LTK were used as the immunogen.
Antigen LTK
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056425

 
novus biologicals canada
NB10056425
Unlabeled 0.1mg Each for $255.80

NB10056425T

 
novus biologicals canada
NB10056425SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 800-864 of human LTK were used as the immunogen.
Antigen LTK
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen has 100% identity in human isoforms 1 and 2, and 81% homologous in mouse isoforms A, B, C, and D, and rat. LTK is an 864 amino acid protein with an approximate molecular weight of 94 kD. Cross reacts with Human. Predicted to cross react with Mouse and Rat.
Gene Accession No. P29376
Gene ID (Entrez) 4058
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL