Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224629SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224629SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 23028
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, and chimpanzee, 94% homologous in rat and cow, 88% homologous in chicken and dog and 83% homologous in zebrafish.
Immunogen A portion of amino acids 450-500 of human LSD1 was used as the immunogen.
Primary or Secondary Primary
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Gene Accession No. O60341
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Format Purified
Quantity 0.025mg
Antigen LSD1
Host Species Rabbit
Applications Simple Western
Applications Western Blotting