Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224629

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224629

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, and chimpanzee, 94% homologous in rat and cow, 88% homologous in chicken and dog and 83% homologous in zebrafish.
Concentration 0.5mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 23028
Format Purified
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Immunogen A portion of amino acids 450-500 of human LSD1 was used as the immunogen.
Regulatory Status RUO
Gene Accession No. O60341
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Applications Simple Western
Applications Western Blotting
Quantity 0.1mg
Host Species Rabbit
Antigen LSD1