Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056496SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056496T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a,CRA _b and CRA _c), rhesus monkey, dog (isoforms 1 and 2), mouse (isoforms 1, 2, 3 and 4) and rat. Cross reacts with Dog, Human, Mouse, Rat and Rhesus Monkey.
Gene Accession No. O60341
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen A portion of amino acids 800-850 of human LSD1 was used as the immunogen.
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Conjugate Unlabeled
Gene ID (Entrez) 23028
Antigen LSD1
Host Species Rabbit
Format Purified
Quantity 0.025mg