Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056496

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056496

  • / Each
 

Description & Specifications

Specifications

Antigen LSD1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. O60341
Host Species Rabbit
Immunogen A portion of amino acids 800-850 of human LSD1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23028
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a,CRA _b and CRA _c), rhesus monkey, dog (isoforms 1 and 2), mouse (isoforms 1, 2, 3 and 4) and rat. Cross reacts with Dog, Human, Mouse, Rat and Rhesus Monkey.
Target Species Canine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat