Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 400-450 of human LSD1 were used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224734

 
novus biologicals canada
NBP224734
Unlabeled 0.1mg Each for $255.80

NBP224734SS

 
novus biologicals canada
NBP224734SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 400-450 of human LSD1 were used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, 84% in cow, 80% in dog and 72% homologous in mouse.
Gene Accession No. O60341
Gene ID (Entrez) 23028
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL