Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224629

 
novus biologicals canada
NBP224629
Unlabeled 0.1mg Each for $255.80

NBP224629SS

 
novus biologicals canada
NBP224629SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, and chimpanzee, 94% homologous in rat and cow, 88% homologous in chicken and dog and 83% homologous in zebrafish.
Gene Accession No. O60341
Gene ID (Entrez) 23028
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL