Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 800-850 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056496

 
novus biologicals canada
NB10056496
Unlabeled 0.1mg Each for $255.80

NB10056496T

 
novus biologicals canada
NB10056496SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 800-850 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a,CRA _b and CRA _c), rhesus monkey, dog (isoforms 1 and 2), mouse (isoforms 1, 2, 3 and 4) and rat. Cross reacts with Dog, Human, Mouse, Rat and Rhesus Monkey.
Gene Accession No. O60341
Gene ID (Entrez) 23028
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL