Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056475

 
novus biologicals canada
NB10056475
Unlabeled 0.1mg Each for $257.41

NB10056475T

 
novus biologicals canada
NB10056475SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human LSD1 was used as the immunogen.
Antigen LSD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform b), mouse, rat, dog, zebrafish and rhesus monkey. Cross reacts with Dog, Human, Mouse, Rat, Rhesus Monkey and Zebrafish.
Gene Accession No. O60341
Gene ID (Entrez) 23028
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL