Please login to your online account to display your discounted pricing

anti-LSD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224734

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224734

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Gene ID (Entrez) 23028
Concentration 0.5mg/mL
Gene Accession No. O60341
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Quantity 0.1mg
Formulation 100μg in 200μL PBS containing 0.05% BSA
Format Purified
Host Species Rabbit
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, 84% in cow, 80% in dog and 72% homologous in mouse.
Antigen LSD1
Regulatory Status RUO
Immunogen A portion of amino acids 400-450 of human LSD1 were used as the immunogen.
Applications Western Blotting