Please login to your online account to display your discounted pricing

anti-LRRFIP1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200 to 250 of human LRRFIP1 was used as immunogen.
Antigen LRRFIP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224672

 
novus biologicals canada
NBP224672
Unlabeled 0.1mg Each for $257.41

NBP224672SS

 
novus biologicals canada
NBP224672SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200 to 250 of human LRRFIP1 was used as immunogen.
Antigen LRRFIP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in rhesus monkey and horse and 83% homologous in Tetradon. The theoretical size of NALP10 is approximately 71kD. IMG-6366A detects a protein at approximately 60kD and the response is completely blocked by the immunizing peptide.The reason for the size discrepency is not known.
Gene ID (Entrez) 9208
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL