Please login to your online account to display your discounted pricing

anti-LRRFIP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224672SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224672SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9208
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in rhesus monkey and horse and 83% homologous in Tetradon. The theoretical size of NALP10 is approximately 71kD. IMG-6366A detects a protein at approximately 60kD and the response is completely blocked by the immunizing peptide.The reason for the size discrepency is not known.
Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Antigen LRRFIP1
Format Purified
Quantity 0.025mg
Cross Reactivity Human
Conjugate Unlabeled
Immunogen A portion of amino acid 200 to 250 of human LRRFIP1 was used as immunogen.
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO