Please login to your online account to display your discounted pricing

anti-LRRFIP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224672

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224672

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Gene ID (Entrez) 9208
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Format Purified
Applications Western Blotting
Immunogen A portion of amino acid 200 to 250 of human LRRFIP1 was used as immunogen.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in rhesus monkey and horse and 83% homologous in Tetradon. The theoretical size of NALP10 is approximately 71kD. IMG-6366A detects a protein at approximately 60kD and the response is completely blocked by the immunizing peptide. The reason for the size discrepency is not known.
Antigen LRRFIP1
Host Species Rabbit
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Human