Please login to your online account to display your discounted pricing

anti-LRRC32/GARP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224664

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224664

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 2615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chicken, cow, dog, mouse, rat and monkey, and 92% homologous in xenopus. FOXO3A is a 673 amino acid protein which is observed at approximately 95 kD in western blot.
Concentration 0.5mg/mL
Applications Flow Cytometry
Applications Western Blotting
Primary or Secondary Primary
Antigen LRRC32/GARP
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Gene Accession No. Q14392
Regulatory Status RUO
Conjugate Unlabeled
Immunogen A portion of amino acids 250-300 of human GARP/LRRC32 was used as the immunogen.
Host Species Rabbit