missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-LRG1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human LRG1 was used as the immunogen.
Antigen LRG1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224603 Novus Biologicals Canada
NBP224603
0.1mg Each for $255.80
NBP224603SS Novus Biologicals Canada
NBP224603SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
LRG1
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, 77% homologous in rhesus monkey, and 55% homologous in mouse.
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acids 300-350 of human LRG1 was used as the immunogen.
Polyclonal
Primary
Western Blot
116844
0.5mg/mL