Please login to your online account to display your discounted pricing

anti-LRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224603SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224603SS

  • / Each
 

Description & Specifications

Specifications

Antigen LRG1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human LRG1 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 116844
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, 77% homologous in rhesus monkey, and 55% homologous in mouse.
Target Species Human
Target Species Murine
Target Species Primate