Please login to your online account to display your discounted pricing

anti-LRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224603

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224603

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, 77% homologous in rhesus monkey, and 55% homologous in mouse.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Applications Western Blotting
Quantity 0.1mg
Antigen LRG1
Gene ID (Entrez) 116844
Concentration 0.5mg/mL
Regulatory Status RUO
Format Purified
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human LRG1 was used as the immunogen.