Please login to your online account to display your discounted pricing

anti-LOX propeptide, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot No. P28300]
Antigen LOX propeptide
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP130327SS

 
novus biologicals canada
NBP130327SS
Unlabeled 0.05mL Each for $139.00

NBP130327

 
novus biologicals canada
NBP130327
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot No. P28300]
Antigen LOX propeptide
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity This antibody recognizes the glycosylated propeptide form (∽35 kDa) and the proenzyme form (∽50 kDa).
Gene Accession No. P28300
Gene ID (Entrez) 4015
Formulation PBS, 30% glycerol
Concentration 1mg/mL