Please login to your online account to display your discounted pricing

anti-LOX propeptide, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155). [Swiss-Prot P28301]
Antigen LOX propeptide
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP130012

 
novus biologicals canada
NBP130012
Unlabeled 0.1mL Each for $309.05

NBP130012SS

 
novus biologicals canada
NBP130012SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155). [Swiss-Prot P28301]
Antigen LOX propeptide
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Simple Western
Applications Western Blotting
Test Specificity Human and mouse. Immunogen has 100% homology with rat.
Gene Accession No. P28300
Gene ID (Entrez) 4015
Formulation PBS
Concentration 1mg/mL