Please login to your online account to display your discounted pricing

anti-LOX propeptide, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130327SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130327SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Test Specificity This antibody recognizes the glycosylated propeptide form (∽35 kDa) and the proenzyme form (∽50 kDa).
Gene ID (Entrez) 4015
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Host Species Rabbit
Antigen LOX propeptide
Gene Accession No. P28300
Concentration 1mg/mL
Immunogen Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot No. P28300]
Quantity 0.05mL
Regulatory Status RUO