Please login to your online account to display your discounted pricing

anti-LOX propeptide, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130327

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130327

  • / Each
 

Description & Specifications

Specifications

Test Specificity This antibody recognizes the glycosylated propeptide form (∽35 kDa) and the proenzyme form (∽50 kDa).
Gene Accession No. P28300
Primary or Secondary Primary
Format Affinity Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Gene ID (Entrez) 4015
Formulation PBS, 30% glycerol
Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Rat
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot No. P28300]
Host Species Rabbit
Antigen LOX propeptide