Please login to your online account to display your discounted pricing

anti-LOX propeptide, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130012

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130012

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. P28300
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4015
Test Specificity Human and mouse. Immunogen has 100% homology with rat.
Conjugate Unlabeled
Antigen LOX propeptide
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Simple Western
Applications Western Blotting
Immunogen A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155). [Swiss-Prot P28301]
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit
Primary or Secondary Primary
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL