Please login to your online account to display your discounted pricing

anti-LOX, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 300-350). [Swiss-Prot P28300]
Antigen LOX
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002528SS

 
novus biologicals canada
NB1002528SS
Unlabeled 0.025mL Each for $109.00

NB1002528

 
novus biologicals canada
NB1002528
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 300-350). [Swiss-Prot P28300]
Antigen LOX
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human and mouse. Predicted to react with rat and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to porcine, Xenopus and Zebrafish.
Gene Accession No. P28300
Gene ID (Entrez) 4015
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL