Please login to your online account to display your discounted pricing

anti-LOX, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11059729

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11059729

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4015
Format Antiserum
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Immunogen Two synthetic peptides (within residues 150-200 and 275-325) of the rat LOX protein. [UniProt No. P16636]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Host Species Rabbit
Antigen LOX