Please login to your online account to display your discounted pricing

anti-LOX, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002528SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002528SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4015
Gene Accession No. P28300
Concentration 1mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 300-350). [Swiss-Prot P28300]
Quantity 0.025mL
Antigen LOX
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Formulation Tris-glycine, 150mM NaCl
Test Specificity Human and mouse. Predicted to react with rat and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to porcine, Xenopus and Zebrafish.
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Format Affinity Purified