Please login to your online account to display your discounted pricing

anti-LOX, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002528

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002528

  • / Each
 

Description & Specifications

Specifications

Antigen LOX
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P28300
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human LOX protein (within residues 300-350). [Swiss-Prot P28300]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4015
Test Specificity Human and mouse. Predicted to react with rat and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to porcine, Xenopus and Zebrafish.
Target Species Human
Target Species Murine