Please login to your online account to display your discounted pricing

anti-LOX, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Two synthetic peptides (within residues 150-200 and 275-325) of the rat LOX protein. [UniProt No. P16636]
Antigen LOX
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11059729T

 
novus biologicals canada
NB11059729SS
Unlabeled 0.025mL Each for $139.00

NB11059729

 
novus biologicals canada
NB11059729
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Two synthetic peptides (within residues 150-200 and 275-325) of the rat LOX protein. [UniProt No. P16636]
Antigen LOX
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 4015