Please login to your online account to display your discounted pricing

anti-LMW-PTP/ACP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056474

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056474

  • / Each
 

Description & Specifications

Specifications

Antigen LMW-PTP/ACP1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_009030
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human ACP1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 52
Test Specificity The amino acid sequence used as the immunogen for this antibody is 100% homologous in human, 93% in dog and orangutan, 86% in chicken and 80% homologous in mouse and rat (isoform a). It is also 100% homologous in human ACP1 isoforms c and d. An uncharacterized 45 kD band is also seen and blocked by the immunizing peptide. Cross reacts with Human. Predicted to react with Mouse, Rat, Dog, Chicken and Orangutan.
Target Species Human