Please login to your online account to display your discounted pricing

anti-LMW-PTP/ACP1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human ACP1 was used as the immunogen.
Antigen LMW-PTP/ACP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056474

 
novus biologicals canada
NB10056474
Unlabeled 0.1mg Each for $255.80

NB10056474T

 
novus biologicals canada
NB10056474SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human ACP1 was used as the immunogen.
Antigen LMW-PTP/ACP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as the immunogen for this antibody is 100% homologous in human, 93% in dog and orangutan, 86% in chicken and 80% homologous in mouse and rat (isoform a). It is also 100% homologous in human ACP1 isoforms c and d. An uncharacterized 45 kD band is also seen and blocked by the immunizing peptide. Cross reacts with Human. Predicted to react with Mouse, Rat, Dog, Chicken and Orangutan.
Gene Accession No. NP_009030
Gene ID (Entrez) 52
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL