Please login to your online account to display your discounted pricing

anti-LMO2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11083978

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11083978

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Conjugate Unlabeled
Gene Accession No. P25791
Test Specificity Human, mouse, rat and bovine. Immunogen sequence has 93% homology to Zebrafish.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Applications Chromatin Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen LMO2
Immunogen Synthetic peptide made to an N-terminal portion of the human LMO2 protein (within residues 1-100). [Swiss-Prot No. P25791]
Gene ID (Entrez) 4005
Concentration 1mg/mL
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO