Please login to your online account to display your discounted pricing

anti-LMO2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11083978SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11083978T

  • / Each
 

Description & Specifications

Specifications

Antigen LMO2
Applications Chromatin Immunoprecipitation
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P25791
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of the human LMO2 protein (within residues 1-100). [Swiss-Prot No. P25791]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4005
Test Specificity Human, mouse, rat and bovine. Immunogen sequence has 93% homology to Zebrafish.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Rat