Please login to your online account to display your discounted pricing

anti-LKB1/STK11, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP214834

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP214834

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Concentration 1.16mg/mL
Gene Accession No. Q15831
Gene ID (Entrez) 6794
Conjugate Unlabeled
Antigen LKB1/STK11
Regulatory Status RUO
Primary or Secondary Primary
Quantity 0.1mL
Immunogen A synthetic peptide made to a N-terminal portion of the human LKB1 protein (between residues 1-100) [UniProt Q15831]
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit