Please login to your online account to display your discounted pricing

anti-LKB1/STK11, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a N-terminal portion of the human LKB1 protein (between residues 1-100) [UniProt Q15831]
Antigen LKB1/STK11
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP214834

 
novus biologicals canada
NBP214834
Unlabeled 0.1mL Each for $309.05

NBP214834SS

 
novus biologicals canada
NBP214834SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a N-terminal portion of the human LKB1 protein (between residues 1-100) [UniProt Q15831]
Antigen LKB1/STK11
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q15831
Gene ID (Entrez) 6794
Formulation PBS, 30% glycerol
Concentration 1.16mg/mL