Please login to your online account to display your discounted pricing

anti-Liprin alpha 1, Polyclonal, Novus Biologicals™

$472.00 - $472.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Liprin alpha 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

21720002

 
novus biologicals canada
21720002
Unlabeled 0.05mg Each for $472.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Liprin alpha 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This product is specific for Human PPFIA1.
Gene Accession No. NM_177423
Gene ID (Entrez) 8500
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0