missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Lipin 1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide to an internal region (within residues 500-600) of the human LPIN1 protein. [Swiss-Prot No. Q14693]
Antigen Lipin 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11057151 Novus Biologicals Canada
NB11057151
0.1mL Each for $329.00
NB11057151T Novus Biologicals Canada
NB11057151SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Lipin 1
Affinity Purified
RUO
Mouse and human. Immunogen has 94% homology to primate and 76% homology to rat protein.
23175
1.08mg/mL
A synthetic peptide to an internal region (within residues 500-600) of the human LPIN1 protein. [Swiss-Prot No. Q14693]
Polyclonal
Primary
Western Blot
Q14693
PBS, 30% glycerol