Please login to your online account to display your discounted pricing

anti-Lipin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11057151SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11057151T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity Mouse and human. Immunogen has 94% homology to primate and 76% homology to rat protein.
Concentration 1.08mg/mL
Primary or Secondary Primary
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide to an internal region (within residues 500-600) of the human LPIN1 protein. [Swiss-Prot No. Q14693]
Applications Western Blotting
Gene ID (Entrez) 23175
Gene Accession No. Q14693
Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen Lipin 1
Format Affinity Purified
Host Species Rabbit